Synonyms:
2-Propylzinc bromide
Packaged under argon in resealable ChemSeal™ bottles.
H58536.AD
H58536.ADEA
267
EUR
InStock
H58536.AD
Isopropylzinc bromide 0.5 M dans le THF
Isopropylzinc bromide
Isopropylzinc bromide is employed in croos-coupling reactions.
Handle under dry protective gas. Air sensitive. Store away from strong bases and oxidizing agents, heat, acids and water.