H63583.30
H63583.30EA
404
EUR
InStock
H63583.30
2,4,6-Tripropyl-2,4,6-trioxo-1,3,5,2,4,6-trioxatriphosphorinane ≥50% (p/p) dans le dichloromethane
2,4,6-Tripropyl-2,4,6-trioxo-1,3,5,2,4,6-trioxatriphosphorinane
CAS No.: 68957-94-8
Molecular Formula: C9H21O6P3
Formula Weight: 318.19
Physical Form: Liquid
Appearance: Colourless to brown
Beilstein Reference: 5079255
UN No.: UN2922
MDL No.: MFCD00006583