H64366.06
H64366.06EA
68.5
EUR
InStock
H64366.06
2,6-Dichloro-3-iodopyridine 98%
2,6-Dichloro-3-iodopyridine
Purity: 98%
CAS No.: 148493-37-2
Molecular Formula: C5H2Cl2IN
Formula Weight: 273.88
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Sensitivity: Light Sensitive
Appearance: White to brown
Melting Point: 74-79°
Beilstein Reference: 6135696
UN No.: UN2811
MDL No.: MFCD03094686