Synonyms:
3-(Aminomethyl)benzoic acid, HCl
H64546.03
H64546.03EA
101
EUR
InStock
H64546.03
3-(Aminomethyl)benzoic acid hydrochloride 95%
3-(Aminomethyl)benzoic acid hydrochloride
Purity: 95%
CAS No.: 876-03-9
Molecular Formula: C8H9NO2.HCl
Formula Weight: 187.62
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Melting Point: 242-244°
Beilstein Reference: 3697656
MDL No.: MFCD03791117