H66262.22
H66262.22EA
277
EUR
InStock
H66262.22
N-Isopropylacrylamide 97%
N-Isopropylacrylamide
Purity: 97%
CAS No.: 2210-25-5
Molecular Formula: C6H11NO
Formula Weight: 113.16
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Melting Point: 60-63°
Boiling Point: 89-92°/2mm
Beilstein Reference: 1744926
MDL No.: MFCD00041913