Waste water treatment plant, portable laboratory, Model STPL-WRT
Fournisseur: Hach
188703
HACH188703EA 1650 EUR InStock
HACH188703
Waste water treatment plant, portable laboratory, Model STPL-WRT
Kits de tests environnementaux
Wastewater treatment plants, water reclamation facilities and industrial pretreatment operations require operators to perform basic process control testing at multiple sites within their facilities. On-site testing and immediate feedback of results help operators optimize and understand changes in the treatment process. Utilizing drop count titration and color disc colorimetry, this kit tests for temperature, pH (4 to 10), dissolved oxygen (1 to 10 mg/L), and chlorine (0 to 3,4 mg/L).
  • Complete, self-contained portable laboratory
  • Contains everything needed for measurements
  • Easy to use
Information de livraison: Includes apparatus and case
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité