Water quality test discs, Model HA-77
Fournisseur: Hach
202300
HACH202300EA 231 EUR InStock
HACH202300
Water quality test discs, Model HA-77
Kits de tests environnementaux
Hardness and iron determination in a water quality test kit which offers accuracy and portable utility in one package. Utilising drop count titration (hardness) and colour disc colorimetry.
  • Easy to use
  • Portable kit
Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accommodate these items.
Information de livraison: Includes four reagents, bottles, flask, color discs, comparator box, viewing tubes, instructions and carrying case sufficient for 100 tests.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité