Test kits, acidity, model AC-DT
Fournisseur: Hach
2064000
HACH2064000EA 592 EUR InStock
HACH2064000
Test kits, acidity, model AC-DT
Kits de tests environnementaux
This kit is used to determine acidity concentrations.
  • Digital titrator offers higher levels of precision and accuracy than drop count procedures
  • Rugged titration device is ideal for field use, with no glass burettes required
Acidity is usually caused by the presence of mineral acids, salts of strong acids or free carbon dioxide. In surface or ground waters, acidity may form industrial pollution which in excessive levels cause corrosion and can be detrimental to fish.
Please note some of the reagents supplied in the kits are hazardous and appropriate precautions should be taken for use and disposal. Please contact Avantor for details.
Information de livraison: Includes digital titrator, reagents and labware, manual and carrying case.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité