Hardness (Ca and total sequential) HR reagent set, digital titrator
Fournisseur: Hach
2448700
HACH2448700EA 168 EUR InStock
HACH2448700
Hardness (Ca and total sequential) HR reagent set, digital titrator
Kits de tests environnementaux
For sequential determination of calcium and total hardness by EDTA digital titrator titration.
  • Accurate results; when and where you need them
  • Fast and flexible
  • Versatile
  • Economical
Hach method 8329. Range: 100 to 4000 mg/L CaCO₃. Set contains buffer solution (Hardness 1), CalVer® 2 calcium indicator powder pillows, ManVer® 2 hardness indicator powder pillows, potassium hydroxide solution (8 N), sulfuric acid (5,25 N) and 0,800 M EDTA digital titrator cartridge. Approximately 40 to 100 tests.
Informations de commande: Hazardous items; items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accomodate these items.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité