Toxtrak™ toxicity reagent set
Fournisseur: Hach
2597200
HACH2597200EA 753 EUR InStock
HACH2597200
Toxtrak™ toxicity reagent set
Kits de tests environnementaux
Use the toxtrak™ toxicity reagent set to analyze your water or wastewater samples for toxicity.
  • The toxtrak™ reagents perform toxicity screens on drinking water, wastewater and natural water
  • Toxicity is compared to a baseline using indigenous biomass or stock bacteria cultures with a range of zero 100% inhibition
  • A colorimetric method is used to determine the toxicity of samples, with analysis done using one of the hach's trusted spectrophotometers
Follows hach toxicity method 10017.
Informations de commande: Set includes total bacteria count media set (15 TBC tubes, 15 individually wrapped sterile pipets), toxtrak™ powder pillows (pk/50), toxtrak™ accelerator solution (15 ml), disposable culture tubes (5 packs; each pk/6), sterile transfer pipets (pk/15) and sodium thiosulfate standard (100 ml, 0,0246N). Zero to 100% inhibition.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité