Single-Port Portable Parallel Analyser (PPA), SL250
Fournisseur: Hach
LPV4439911002
HACHLPV4439911002EA 2550 EUR InStock
HACHLPV4439911002
Single-Port Portable Parallel Analyser (PPA), SL250
Colorimètres Colorimètres multiparamètres
One portable instrument - all the key parameters.
  • Probe-based tests include pH, dissolved oxygen, conductivity, and more
  • 5-Pin input connector: Two M12 connectors for IntelliCal™ probes
  • 130,8×58,9×258,3 mm dimension
Hach's SL250 Portable parallel analyser (PPA) is an innovative multiparameter water quality meter that allows you to simultaneously perform colorimetric and probe-based measurements. The instrument and corresponding Chemkey reagents are easy to use regardless of technical expertise. Compared to traditional powder or liquid reagent methods, Chemkeys use a simplified testing method that requires no sample preparation, making the process significantly easier, faster, and safer while drastically reducing variability in results.
Colorimetric tests include free chlorine, total chlorine, fluoride, high range manganese, zinc, copper, nitrite, dissolved iron, high and low range alkalinity, high and low range orthophosphate, high and low range hardness, peracetic acid, and more.
Certifications: UKCA, FCC, ISED, RCM, KC.
Information de livraison: SL250 PPA with carry case, 125 V power cable and manual resources.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité