Cuves, rondes
Fournisseur: Hach
LZM381
HACHLZM381EA 0 EUR InStock
HACHLZM381
Cuves, rondes
Cuvettes Cuves de spectrophotométrie
New disposable sample cuvette with screw cap for easy and safe colour measurements. The cuvette is specially designed for the measurement of volatile, toxic or carcinogenic substances. The easily sealable rectangular sample cuvette with screw cap meets the safety requirements of the chemical and pharmazeutical industry.
  • Secure and reliable measurement results because of high beam path lengths
  • Safe handling since there the is no spillage risk due to a closed sample cuvette
  • Protection against outgassing or premature evaporation of the sample liquid
The samples in the sealed cuvette can be directly used as retained samples or stored for long-term ageing tests.
The sealable cuvette is suitable for all Lico colour applications as direct replacement for the existing 50 mm plastic cuvette LZM130, without any verification effort because material and dimensions are exactly the same.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité