Ion selective electrode, sodium, sensION+ 9650
Fournisseur: Hach
LZW9650970002
HACHLZW9650970002EA 1169 EUR InStock
HACHLZW9650970002
Ion selective electrode, sodium, sensION+ 9650
Électrodes Électrodes ISE
sensION+ 9650 is a half-cell sodium ion selective electrode (ISE) with glass body and glass membrane selective to sodium ions in solution. It is recommended for use with the sensION+ 5044 reference electrode.
  • With fixed 1 meter cable and BNC connector
  • For use with a reference electorode
  • Provides fast, stable, and accurate response in a variety of sample types with low maintenance
This electrode is ideal for measuring sodium concentrations in wastewater, drinking water and general water quality applications.
Information de livraison: Supplied with basic user manual.
Commander maintenant
COMMANDE
ACCESSOIRES
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité