Ion selective electrode, half-cell, chloride, sensION+ 9652
Fournisseur: Hach
LZW9652970002
HACHLZW9652970002EA 1169 EUR InStock
HACHLZW9652970002
Ion selective electrode, half-cell, chloride, sensION+ 9652
Électrodes Électrodes ISE
sensION+ 9652 is a half-cell (reference not integrated) chloride ion selective electrode with an epoxy body and non-replaceable crystalline membrane selective to chloride ions in solution.
  • Utilises a unique solid-state sensor technology that eliminates costly membrane replacements
  • Requires virtually no maintenance: Solid-state sensor design allows for dry storage of the ISE without a shelf life or membrane replacement
  • Provides fast, stable, and accurate response in a variety of sample types
The 9652 has a fixed 1 meter cable and BNC connector for laboratory use with the Hach sensION+ MM374 Multimeter. The 9652 is ideal for measuring chloride concentrations in wastewater, drinking water and general water quality applications.
Information de livraison: Supplied with basic user manual.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité