Syringes, SampleLock™
Fournisseur: HAMILTON BONADUZ
81656 81056 86326 86336 86346 81256 81356 81456 81556 80956 81156 CAL81156
HAMI81656EA 387 EUR InStock
HAMI81656 HAMI81056 HAMI86326 HAMI86336 HAMI86346 HAMI81256 HAMI81356 HAMI81456 HAMI81556 HAMI80956 HAMI81156 HAMICAL81156
Syringes, SampleLock™
Seringues Seringues de chromatographie
These syringes allow the collection, storage, transport and analysis of liquid or gaseous samples without the danger of evaporative loss or environmental contamination.
  • Not autoclavable, gas sterilisable
  • An easy-to-use twist valve
  • A positive rear stop prevents loss of sample and plunger blowout
  • Fluid path includes SST, borosilicate glass and PTFE
The sample lock termination incorporates an on/off syringe valve with removable needle. This termination is used for headspace, environmental sample collection and storage, pre-pressurization of gaseous samples for GC analysis, and sample spiking.
Optional male luer and female luer lock adapters (sold separately) threads onto the SampleLock™ valve, making the syringes compatible with a multitude of connectors and fittings.
Commander maintenant
COMMANDE
ACCESSOIRES
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité