Adjustable multi-channel gel loading syringe
Fournisseur: HAMILTON BONADUZ
84611
HAMI84611EA 2680 EUR InStock
HAMI84611
Adjustable multi-channel gel loading syringe
Seringues Seringues de chromatographie
Adjustable multi-channel gel loading syringes simplify and speed sample loading of 96 lane gels. The adjustable gel loading syringe (AGLS) allows quick, easy transfer and loading of DNA samples. Eight samples at a time can be transferred directly from a microwell plate with 9 mm spacing to a 96-lane gel with 10,8 mm spacing.
  • Compatible with 96-lane ABI 377 PRISM® DNA sequencing instrument
  • Easy-to-use design for needle spacing adjustment
Gastight® syringes are ideal for dispensing both liquids and gases. They have a precision machined PTFE plunger tip which creates a leak-free seal.
Removable needle (RN): The design allows the needles to seat precisely at the zero graduation mark of the syringe. Users can select the needle gauge, length and point style to optimise the syringe for custom applications. Additionally, this termination allows for a removable needle without increasing the dead volume of the syringe and is ideal when there is a risk of the needle clogging. Autoclavable when disassembled. Repeated autoclaving will shorten syringe life.
Needle point style 3: Blunt needle point for use with HPLC injection valves and for sample pipetting.
Commander maintenant
COMMANDE
ACCESSOIRES
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité