Technical pH buffer solutions in sachets, HI 50000 series
Fournisseur: Hanna
HI50001-02 HI50003-02 HI50004-02
HANNHI50001-02EA 55.3 EUR InStock
HANNHI50001-02 HANNHI50003-02 HANNHI50004-02
Technical pH buffer solutions in sachets, HI 50000 series
Tampons Tampons pH
These technical calibration buffers produced in accordance with ISO 3696/BS3978 standards using high purity salts, deionised water, certified weight-checked balances, and Class A glassware in a temperature controlled environment monitored with certified thermometers.
  • Technical buffer solutions with certificate
  • Accuracy of ±0,01 pH at 25 °C
  • Colour coded label for easy pH buffer value identification
Temperature chart of the actual pH value at various temperatures are printed on each sachets. Solutions are free from any dyes that may stain the reference cell of a pH electrode.
Certifications: Traceable to NIST Standard Reference Materials (SRMs).
Information de livraison: These solutions are supplied with a certificate of analysis.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité