Test tube racks, OneRack®
Fournisseur: HEATHROW
HD27515A
HEATHD27515AEA 352 EUR InStock
HEATHD27515A
Test tube racks, OneRack®
Portoirs Tube Racks
These durable racks are molded in a single, continuous piece so no assembly required and there are no detachable pieces.
  • Offers the highest chemical and heat resistance (–40 to +121 °C)
  • Have molded coordinates for tube orientation
  • Stable design for use in water baths
  • Flat labeling area for easier identification
  • Unique tear drop shaped openings for better retention of tubes
  • Autoclavable
Made of fiberglass-reinforced polyoxymethylene, these racks are tough and more highly resistant to acids, bases, solvents and heat.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité