Face shields, medical Assistant®
Fournisseur: GLASWARENFABRIK KARL HECHT
49075010
HECH49075010EA 3.5 EUR InStock
HECH49075010
Face shields, medical Assistant®
Écrans de protection
This disposable face shield consists of a headband band made of non allergenic fleece and a transparent PC visor. The face shield covers the entire face, not just the area around the eyes (for example, like safety glasses). This considerably reduces the risk of infection. It is ideal for use in orthopaedics, during general operations, in surgery, urology and dentistry. The shield can also be used by emergency services, as well as in laboratories and emergency wards.
  • Can be worn together with safety glasses
  • Protective film prevents scratches prior to use
  • The moisture-absorbing material of the strap adequately absorbs any perspiration on the brow or temples
  • Velcro fastening for a custom fit
  • Large field of view
  • Prevents direct inhalation of smoke when using a cauter
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité