Chillers, Hei-CHILL 400 Pro
Fournisseur: Heidolph Instruments GmbH & Co.KG
591-01640-00
HEID591-01640-00EA 3650 EUR InStock
HEID591-01640-00
Chillers, Hei-CHILL 400 Pro
Thermostats à circulation Systèmes à circulation fermée Circulateurs de refroidissement
This space-saving model with a cooling capacity of 400 W is ideal for a large number of evaporation tasks in the laboratory.
  • Sustainable and environmentally friendly using natural refrigerants (R-290) and an optimized control
  • Low operating noises for a trouble-free work
RS-232 interface for activating your device via a Hei-VAP expert control or ultimate control or via the Heidolph laboratory software solutions.
Compact light weight for a flexible positioning on the laboratory bench, in a fume hood or, due to its increased pumping capacity, also under the laboratory bench.
Protection class: IP 20
Acoustic pressure: 47 dB
Control accuracy: at −10 °C: ±1 K
Display resolution: 0,1 K
Hose connections: ¹/₄" female thread
Coolant tank: 1,5 to 4,0 L
Max. operating pressure: 25 bar
Working temperature range: −10 up to +40 °C
Permitted ambient temperature range: 5 to 40 °C
Max. temperature of coolant: 60 °C
Circulation pump: Flow rate [max.]: 6 L/min; Flow pressure [max.]: 0,3 bar
Commander maintenant
COMMANDE
SPÉCIFICATIONS
ACCESSOIRES
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité