Chillers, Hei-CHILL 700 Pro
Fournisseur: Heidolph Instruments GmbH & Co.KG
591-01650-00
HEID591-01650-00EA 4473 EUR InStock
HEID591-01650-00
Chillers, Hei-CHILL 700 Pro
Thermostats à circulation Systèmes à circulation fermée Circulateurs de refroidissement
Powerful 700 W cooling for every evaporation process and large volumes.
  • High performance for achieving the target temperature quickly and for keeping values constant even when using high-polar solvents
  • Environmentally friendly by using natural refrigerants (R-290) and an efficient cooling system with low-energy fans
  • Touchscreen controller with glass surface and integrated level indicator
Reduced running noises for a comfortable work thanks to speed-controlled fans in line with your requirements. For applications from −10 to +40 °C with a temperature constancy of ±0,5 °C, reliable cooling for different workflows.
RS-232 connection for activating your device via a Hei-VAP expert control or ultimate control or via the Heidolph laboratory software solutions.
Protection class (EN 60529): IP 20
Acoustic pressure: 49 dB(A)
Hose connections: ⅜" female thread
Coolant tank: 5,5 to 9,0 L
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité