Peltier Circulator, Hei-CHILL
Fournisseur: Heidolph Instruments GmbH & Co.KG
591-01910-00
HEID591-01910-00EA 4990 EUR InStock
HEID591-01910-00
Peltier Circulator, Hei-CHILL
Thermostats à circulation Systèmes à circulation fermée Circulateurs de refroidissement
With a high cooling capacity of 600 W and a heating power being almost twice as high, the Hei-CHILL Peltier thermostat is suitable for a large number of temperature control tasks.
  • 6 L tank volume for temperature stability, even under sustained load
  • Precise temperature regulation and stability of ≤0,05 °C
  • Operating temperature range of −10 to +70 °C
  • Modern 2.83 in. touch display
  • Ethernet, RS 232 and RS 485 interfaces for the integration in processing solutions
The thermostat can be integrated in processes by means of the Hei-PROCESS solutions using different interfaces. Furthermore, the device can be directly controlled via a Hei-VAP Expert Control or Ultimate Control rotary evaporator as recirculation chiller or can be used independently by the integrated touch display.
Electrical: 100/230 V, 50/60 Hz
Condenser type: Air-cooled
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité