Hei-VOLUME Distimatic Pro Industrial Automatic Module
Fournisseur: Heidolph Instruments GmbH & Co.KG
591-61210-00
HEID591-61210-00EA 0 EUR InStock
HEID591-61210-00
Hei-VOLUME Distimatic Pro Industrial Automatic Module
Évaporateurs Évaporateurs rotatifs
For high-performance distillation with the best separation performance, suitable for most solvents (A1 version), also when processing foaming media (A2 version): Version with adapter for Hei-VAP Industrial with glassware of the A series (A1, A2).
  • Sensor- or time-controlled filling of the rotary flask via the removable 7" touchscreen control panel
  • Start automatic cleaning mode and manual operation
  • The emptying time of the coated collector vessel with condensate cooling by the compressor can also be programmed here
  • Operating temperature: 5 to 31 °C at up to 80% rel. humidity; 32 to 40 °C at up to 50% rel. humidity (decreasing linearly)
The control box with the integrated compressor of the Hei-VOLUME Distimatic Pro can be mounted directly with all components on a Hei-VAP Industrial large-scale rotary evaporator using the bracket supplied.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité