Continuous Flow Rotor, TCF-20
Fournisseur: Thermo Scientific
75003012
HERA75003012EA 62370 EUR InStock
HERA75003012
Continuous Flow Rotor, TCF-20
Centrifugeuses Rotors
Perform bioprocessing or separation of large volumes with very low solids content using the Thermo Scientific™ TCF-20 Continuous Flow Rotor. Rotor innovations-from Thermo Scientific™ Auto-Lock™ rotor exchange to Thermo Scientific™ Auto-ID™ instant rotor identification and speed handles on each rotor lid-shorten run set-up time while providing peace-of-mind the rotor is secure.
  • Secure and push-button rotor exchange in less than three seconds
  • Improved safety and convenience
  • No tools required
  • Quickly change rotors and applications
  • Immediate identification of a rotor
  • Rotor specifications automatically loaded into centrifuge parameters
  • Shortens run set-up time and improves safety
  • Accelerates and simplifies rotor lid tightening ensuring lid is properly attached
  • Easier and safer lifting and carrying of rotors further enhanced with the lightweight design
K factor: 826
Biocontainment: Yes
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité