Heratherm® Stability Chamber
Fournisseur: Thermo Scientific
ESS750
HERAESS750EA 0 EUR InStock
HERAESS750
Heratherm® Stability Chamber
Chambres d'essai Chambres d'humidification
Involved temperature and humidity controls, well suited for stability testing, shelf life testing, and environmental testing.
  • Broad relative humidity range to cover many applications, including humidity levels below ambient
  • Environmentally friendly water-efficient humidification system
  • Minimal condensate in standard applications
  • Integrated water tank, fixed water connection for in-house purified water option available
  • Data retrieval via USB stick, Ethernet connection, or Thermo Cloud via OPC-UA standard
  • Intuitive touchscreen interface with easy programmability of temperature cycles and process duration
  • Simplified setup with no technician required, install using standard power cord
  • Inner glass door keeps environment stable, even when opening outer door
  • Rounded corners and removable shelving offer easy cleaning to help prevent cross-contamination
Energy-efficient Peltier technology supports temperature stability and uniformity with lower energy usage compared to traditional compressors and without hazardous refrigerant. Two access ports for independent data capture, and internal documentation of set and actual parameter values, along with event log to meet 21 CFR Part 11 requirements for process documentation.
Sample and process protection: Assign personalised access with user IDs; restrict access with standard door lock, or optional automated door lock; various alarm options to prevent operator of process issues.
Duplex interior electrical sockets for use of stirrers, shakers or other equipment inside the chamber are available as a factory-installed option. Choose from a variety of country-specific sockets. (Ordered with separate order code).
Data outputs: USB, Ethernet
Chamber material: Powder-coated cold rolled steel
Plug type: Nema 5-20P
Temperature range: 0 to 70 °C
Display: 7" touchscreen user interface
Ports: 2 access ports
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité