SuperNuova+™ Multi-Position Magnetic Stirrers
Fournisseur: Thermo Fisher Scientific
SP135930-33Q S136030-33Q
BARNSP135930-33QEA 2430 EUR InStock
BARNSP135930-33Q HERAS136030-33Q
SuperNuova+™ Multi-Position Magnetic Stirrers
Agitateurs Agitateurs magnétiques multipostes
Carry out solubility studies, digestion, titration and enzymatic studies with Thermo Scientific™ Super-Nuova™ Multi-Position Digital Stirrers. Offering four individually-controlled stirring positions, ceramic tops and 4-function memory, these stirrers are designed for stronger coupling and stirring control with advanced safety features. Ideal for labs with multiple users.
  • Thermo Scientific™ Stir Trac™ stir control provides improved slow-speed stirring, consistent speed, and strong magnetic coupling - reducing the risk of stir bar decoupling
  • Rugged low-profile cast-aluminum body provides stability and durability
  • Microprocessor control ensures accurate and stable setpoints for stirring speed
  • Stir Trac™ braking feature brings stir bar to an immediate stop for quick flask removal
  • Clean easily and resist alkalis and acids
  • Single control knob controls speed
  • Lock feature prevents accidental changes
  • Digital rpm display
  • Seamless
  • Reflective white ceramic surface aids sample visibility
  • Stirring adjustable in units of 1 rpm
  • RS232 port outputs time count
  • Stir speed (set point) directly to your PC
  • Rugged low-profile cast-aluminum body provides stability and durability
  • Unit design protects internal components from accidental spills
  • Walk-away timer shuts off stirring after 1, 2, 4, 8, or 12 hr
  • Detachable line cord and plug
Certifications: CCSAus, CE.
Information de livraison: Supplied with detachable line cord and plug.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité