J&W GS-OxyPLOT GC columns
Fournisseur: Agilent
115-4912E
HEWL115-4912EEA 2700 EUR InStock
HEWL115-4912E
J&W GS-OxyPLOT GC columns
Colonnes pour chromatographie
GS-OxyPlot provides strong selectivity for a wide range of oxygenates and accurate analysis to ppm/ppb levels.
  • Accurate analysis of ppm/ppb level oxygenates in C1 - C10 hydrocarbons
  • Strong selectivity for a wide range of oxygenates (ethers, alcohols, aldehydes, and ketones) in complexmatrices such as gaseous hydrocarbons, motor fuels, and crude oil
  • Suitable for ASTM methods for oxygenates
  • Very high column stability (upper temperature limit of 350 °C) with no column bleed
  • Stable phase coating virtually eliminating particle generation and detector spiking
J&W GS-OxyPLOT is suitable for ASTM methods for oxygenates and is a good alternative to GC columns with gas-liquid phases such as TCEP that are often used for similar applications. GS-OxyPLOT has an upper temperature limit of 350 °C and exhibits none of the bleed that is normally associated with liquid- and siloxane-coated GC columns. With our advanced dynamic coating process, the GS-OxyPLOT stationary phase exhibits excellent stability with virtually no detector spiking due to stationary phase particle shedding.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité