Agilent J&W DB-1301 GC column, 20 m, 0.18 mm, 2.0 um, 7 inch cage. Specifications:- Capillary Column Phase: DB-1301. Capillary Tubin g: Fused Silica. Film Thickness: 2 um. GC Column Format: 7 inch. Inner Diameter (ID): 0.18 mm. Length: 20 m. Length ( 1 * 1 Pce
Fournisseur: Agilent
Agilent J&W DB-1301 GC column, 20 m, 0.18 mm, 2.0 um, 7 inch cage. Specifications:- Capillary Column Phase: DB-1301. Capillary Tubin g: Fused Silica. Film Thickness: 2 um. GC Column Format: 7 inch. Inner Diameter (ID): 0.18 mm. Length: 20 m. Length ( 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Agilent J&W DB-1301 GC column, 20 m, 0.18 mm, 2.0 um, 7 inch cage. Specifications:- Capillary Column Phase: DB-1301. Capillary Tubin g: Fused Silica. Film Thickness: 2 um. GC Column Format: 7 inch. Inner Diameter (ID): 0.18 mm. Length: 20 m. Length ( 1 * 1 Pce HEWL121-1325 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou