Specifications:- Capillary Column Phase: DB-5ht. Capillary Tubing: High Temperature Fused Silica. Film Thickness: 0.18 um. GC Column Format: 7 inch. Inner Diameter (ID): 0.18 mm. Length: 20 m. Length (m): 20 m. Polarity: Low Polarity. With Smart Key 1 * 1 Pce
Fournisseur: Agilent
Specifications:- Capillary Column Phase: DB-5ht. Capillary Tubing: High Temperature Fused Silica. Film Thickness: 0.18 um. GC Column Format: 7 inch. Inner Diameter (ID): 0.18 mm. Length: 20 m. Length (m): 20 m. Polarity: Low Polarity. With Smart Key 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- Capillary Column Phase: DB-5ht. Capillary Tubing: High Temperature Fused Silica. Film Thickness: 0.18 um. GC Column Format: 7 inch. Inner Diameter (ID): 0.18 mm. Length: 20 m. Length (m): 20 m. Polarity: Low Polarity. With Smart Key 1 * 1 Pce HEWL121-5722 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou