Specifications:- Capillary Column Phase: DB-WAX Ultra Inert. Capillary Tubing: Fused Silica. Film Thickness: 0.18 um. GC Column Form at: 7 inch. GC Column Type: Ultra Inert WAX. Inner Diameter (ID): 0.18 mm. Length: 40 m. 1 * 1 Pce
Fournisseur: Agilent
Specifications:- Capillary Column Phase: DB-WAX Ultra Inert. Capillary Tubing: Fused Silica. Film Thickness: 0.18 um. GC Column Form at: 7 inch. GC Column Type: Ultra Inert WAX. Inner Diameter (ID): 0.18 mm. Length: 40 m. 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- Capillary Column Phase: DB-WAX Ultra Inert. Capillary Tubing: Fused Silica. Film Thickness: 0.18 um. GC Column Form at: 7 inch. GC Column Type: Ultra Inert WAX. Inner Diameter (ID): 0.18 mm. Length: 40 m. 1 * 1 Pce HEWL121-7042UI 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou