Specifications:- Capillary Column Phase: DB-624 Ultra Inert. Capillary Tubing: Fused Silica. Film Thickness: 1.4 um. GC Column Forma t: Intuvo. GC Column Type: Ultra Inert. Inner Diameter (ID): 0.25 mm. Length: 60 m. Polarity: Mid Polarity. With Smar 1 * 1 Pce
Fournisseur: Agilent
Specifications:- Capillary Column Phase: DB-624 Ultra Inert. Capillary Tubing: Fused Silica. Film Thickness: 1.4 um. GC Column Forma t: Intuvo. GC Column Type: Ultra Inert. Inner Diameter (ID): 0.25 mm. Length: 60 m. Polarity: Mid Polarity. With Smar 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- Capillary Column Phase: DB-624 Ultra Inert. Capillary Tubing: Fused Silica. Film Thickness: 1.4 um. GC Column Forma t: Intuvo. GC Column Type: Ultra Inert. Inner Diameter (ID): 0.25 mm. Length: 60 m. Polarity: Mid Polarity. With Smar 1 * 1 Pce HEWL122-1364UI-INT 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou