Specifications:- Capillary Column Phase: DB-35ms. Capillary Tubing: Fused Silica. Film Thickness: 0.25 um. GC Column Format: 7 inch. GC Column Type: GC/MS. Inner Diameter (ID): 0.25 mm. Length: 30 m. Polarity: Mid Polarity. With Smart Key: Yes. 1 * 1 Pce
Fournisseur: Agilent
Specifications:- Capillary Column Phase: DB-35ms. Capillary Tubing: Fused Silica. Film Thickness: 0.25 um. GC Column Format: 7 inch. GC Column Type: GC/MS. Inner Diameter (ID): 0.25 mm. Length: 30 m. Polarity: Mid Polarity. With Smart Key: Yes. 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- Capillary Column Phase: DB-35ms. Capillary Tubing: Fused Silica. Film Thickness: 0.25 um. GC Column Format: 7 inch. GC Column Type: GC/MS. Inner Diameter (ID): 0.25 mm. Length: 30 m. Polarity: Mid Polarity. With Smart Key: Yes. 1 * 1 Pce HEWL122-3832-KEY 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou