Specifications:- Capillary Column Phase: DB-WAX Ultra Inert. Capillary Tubing: Fused Silica. Film Thickness: 0.25 um. GC Column Form at: Intuvo. GC Column Type: Ultra Inert WAX. Inner Diameter (ID): 0.25 mm. Length: 5 m. 1 * 1 Pce
Fournisseur: Agilent
Specifications:- Capillary Column Phase: DB-WAX Ultra Inert. Capillary Tubing: Fused Silica. Film Thickness: 0.25 um. GC Column Form at: Intuvo. GC Column Type: Ultra Inert WAX. Inner Diameter (ID): 0.25 mm. Length: 5 m. 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- Capillary Column Phase: DB-WAX Ultra Inert. Capillary Tubing: Fused Silica. Film Thickness: 0.25 um. GC Column Form at: Intuvo. GC Column Type: Ultra Inert WAX. Inner Diameter (ID): 0.25 mm. Length: 5 m. 1 * 1 Pce HEWL122-7002UI-INT 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou