Agilent J&W DB-EUPAH GC column, 15 m, 0.25 mm, 0.25 um. Specifications:- Capillary Column Phase: DB-EUPAH. Capillary Tubing: Fused S ilica. Film Thickness: 0.25 um. GC Column Format: 7 inch. Inner Diameter (ID): 0.25 mm. Length: 15 m. Length (m): 15 1 * 1 Pce
Fournisseur: Agilent
Agilent J&W DB-EUPAH GC column, 15 m, 0.25 mm, 0.25 um. Specifications:- Capillary Column Phase: DB-EUPAH. Capillary Tubing: Fused S ilica. Film Thickness: 0.25 um. GC Column Format: 7 inch. Inner Diameter (ID): 0.25 mm. Length: 15 m. Length (m): 15 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Agilent J&W DB-EUPAH GC column, 15 m, 0.25 mm, 0.25 um. Specifications:- Capillary Column Phase: DB-EUPAH. Capillary Tubing: Fused S ilica. Film Thickness: 0.25 um. GC Column Format: 7 inch. Inner Diameter (ID): 0.25 mm. Length: 15 m. Length (m): 15 1 * 1 Pce HEWL122-9612 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou