Specifications:- Capillary Column Phase: DB-8270D Ultra Inert. Capillary Tubing: Fused Silica. Film Thickness: 0.25 um. GC Column Fo rmat: Intuvo. GC Column Type: Ultra Inert. Inner Diameter (ID): 0.25 mm. Length: 30 m. With Smart Key: Yes. 1 * 1 Pce
Fournisseur: Agilent
Specifications:- Capillary Column Phase: DB-8270D Ultra Inert. Capillary Tubing: Fused Silica. Film Thickness: 0.25 um. GC Column Fo rmat: Intuvo. GC Column Type: Ultra Inert. Inner Diameter (ID): 0.25 mm. Length: 30 m. With Smart Key: Yes. 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- Capillary Column Phase: DB-8270D Ultra Inert. Capillary Tubing: Fused Silica. Film Thickness: 0.25 um. GC Column Fo rmat: Intuvo. GC Column Type: Ultra Inert. Inner Diameter (ID): 0.25 mm. Length: 30 m. With Smart Key: Yes. 1 * 1 Pce HEWL122-9732-INT 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou