DB-TPH GC columns, J&W
Fournisseur: Agilent
123-1632
HEWL123-1632EA 872 EUR InStock
HEWL123-1632
DB-TPH GC columns, J&W
Colonnes pour chromatographie
DB-TPH is specifically designed for the analysis of total petroleum hydrocarbons (TPH), soil analysis, and leaking underground fuel tanks (LUFT).
  • Specifically designed for the analysis of total petroleum hydrocarbons (TPHs), soil analysis, and LUFT
  • Three analyses in one injection – gas range organics, diesel range organics and motor oil
  • Fast run time
  • Bonded and cross-linked
  • Solvent rinsable
DB-TPH columns feature three analyses in a single injection (gas range organics, diesel range organics, and motor oil) and are fast run time columns. With solvent rinsable functionality and an operating temperature range of −10 to +320 °C, these high quality and extensively QC tested columns are excellent for petroleum analysis applications.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité