Specifications:- Capillary Column Phase: DB-5ht. Capillary Tubing: High Temperature Fused Silica. Film Thickness: 0.1 um. GC Column Format: Intuvo. Inner Diameter (ID): 0.32 mm. Length: 5 m. Polarity: Low Polarity. With Smart Key: Yes. 1 * 1 Pce
Fournisseur: Agilent
Specifications:- Capillary Column Phase: DB-5ht. Capillary Tubing: High Temperature Fused Silica. Film Thickness: 0.1 um. GC Column Format: Intuvo. Inner Diameter (ID): 0.32 mm. Length: 5 m. Polarity: Low Polarity. With Smart Key: Yes. 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- Capillary Column Phase: DB-5ht. Capillary Tubing: High Temperature Fused Silica. Film Thickness: 0.1 um. GC Column Format: Intuvo. Inner Diameter (ID): 0.32 mm. Length: 5 m. Polarity: Low Polarity. With Smart Key: Yes. 1 * 1 Pce HEWL123-57J1-INT 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou