Specifications:- Capillary Column Phase: DB-WAX. Capillary Tubing: Fused Silica. Film Thickness: 0.10 um. GC Column Format: Intuvo. GC Column Type: WAX. Inner Diameter (ID): 0.10 mm. Length: 10 m. 1 * 1 Pce
Fournisseur: Agilent
Specifications:- Capillary Column Phase: DB-WAX. Capillary Tubing: Fused Silica. Film Thickness: 0.10 um. GC Column Format: Intuvo. GC Column Type: WAX. Inner Diameter (ID): 0.10 mm. Length: 10 m. 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- Capillary Column Phase: DB-WAX. Capillary Tubing: Fused Silica. Film Thickness: 0.10 um. GC Column Format: Intuvo. GC Column Type: WAX. Inner Diameter (ID): 0.10 mm. Length: 10 m. 1 * 1 Pce HEWL127-7012-INT 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou