PSD spares and supplies
Fournisseur: VARIAN
6610025900 9910091200 9910115100 1510146800
VARI6610025900EA 234 EUR InStock
VARI6610025900 VARI9910091200 VARI9910115100 HEWL1510146800
PSD spares and supplies
Matériel de chromatographie Chromatographie, Pièces de rechange
The Programmable Sample Dispensers (PSD) are high-capacity autosamplers that are used with Graphite Furnace AA.
  • Plug and play mounting with an ultrastable mounting and position lock, eliminating re-alignment
  • Provides capacity for up to up to 50 samples in disposable 2 ml microvials, plus up to five central vessels for blank, bulk standard, and chemical modifiers
  • Can automatically perform sample preparation processes for GFAAS, eliminating time-consuming manual sample preparation
  • Provides flexible dispensing options that are commonly used in GFAAS, including hot inject, multiple injection, and pre- or postaddition of modifiers
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité