Specifications:- Capillary Column Phase: HP-PLOT Q. Capillary Tubing: Fused Silica. Film Thickness: 20 um. GC Column Format: Intuvo. GC Column Type: PLOT. Inner Diameter (ID): 0.32 mm. Length: 15 m. Length (m): 15 m. With Smart Key: Yes. 1 * 1 Pce
Fournisseur: Agilent
Specifications:- Capillary Column Phase: HP-PLOT Q. Capillary Tubing: Fused Silica. Film Thickness: 20 um. GC Column Format: Intuvo. GC Column Type: PLOT. Inner Diameter (ID): 0.32 mm. Length: 15 m. Length (m): 15 m. With Smart Key: Yes. 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- Capillary Column Phase: HP-PLOT Q. Capillary Tubing: Fused Silica. Film Thickness: 20 um. GC Column Format: Intuvo. GC Column Type: PLOT. Inner Diameter (ID): 0.32 mm. Length: 15 m. Length (m): 15 m. With Smart Key: Yes. 1 * 1 Pce HEWL19091P-QO3PT-I 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou