Specifications:- Capillary Column Phase: HP-1ms. Capillary Tubing: Fused Silica. Film Thickness: 0.18 um. GC Column Format: Intuvo. GC Column Type: GC/MS. Inner Diameter (ID): 0.18 mm. Length: 20 m. Polarity: Low Polarity. With Smart Key: Yes. 1 * 1 Pce
Fournisseur: Agilent
Specifications:- Capillary Column Phase: HP-1ms. Capillary Tubing: Fused Silica. Film Thickness: 0.18 um. GC Column Format: Intuvo. GC Column Type: GC/MS. Inner Diameter (ID): 0.18 mm. Length: 20 m. Polarity: Low Polarity. With Smart Key: Yes. 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- Capillary Column Phase: HP-1ms. Capillary Tubing: Fused Silica. Film Thickness: 0.18 um. GC Column Format: Intuvo. GC Column Type: GC/MS. Inner Diameter (ID): 0.18 mm. Length: 20 m. Polarity: Low Polarity. With Smart Key: Yes. 1 * 1 Pce HEWL19091S-677UI-I 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou