Specifications:- Capillary Column Phase: HP-INNOWax. Capillary Tubing: Fused Silica. Film Thickness: 1 um. GC Column Format: 7 inch. Inner Diameter (ID): 0.53 mm. Length: 15 m. Polarity: High Polarity. With Smart Key: No. 1 * 1 Pce
Fournisseur: Agilent
Specifications:- Capillary Column Phase: HP-INNOWax. Capillary Tubing: Fused Silica. Film Thickness: 1 um. GC Column Format: 7 inch. Inner Diameter (ID): 0.53 mm. Length: 15 m. Polarity: High Polarity. With Smart Key: No. 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- Capillary Column Phase: HP-INNOWax. Capillary Tubing: Fused Silica. Film Thickness: 1 um. GC Column Format: 7 inch. Inner Diameter (ID): 0.53 mm. Length: 15 m. Polarity: High Polarity. With Smart Key: No. 1 * 1 Pce HEWL19095N-121I 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou