Specifications:- Chemical Compatibility: Aqueous Organic solvents. Compatible with Advanced Valve System (AVS): Yes. Nebulizer Gas C onnector: Ezylok. Nebulizer Material: Borosilicate glass. Nebulizer Name: MicroMist. Nebulizer Sample Particle Size T 1 * 1 Pce
Fournisseur: Agilent
Specifications:- Chemical Compatibility: Aqueous Organic solvents. Compatible with Advanced Valve System (AVS): Yes. Nebulizer Gas C onnector: Ezylok. Nebulizer Material: Borosilicate glass. Nebulizer Name: MicroMist. Nebulizer Sample Particle Size T 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- Chemical Compatibility: Aqueous Organic solvents. Compatible with Advanced Valve System (AVS): Yes. Nebulizer Gas C onnector: Ezylok. Nebulizer Material: Borosilicate glass. Nebulizer Name: MicroMist. Nebulizer Sample Particle Size T 1 * 1 Pce HEWL2010126000 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou