Specifications:- LC Application Compatibility: Analytical. LC Module Type Compatibility: Autosampler Analytical. LC Supplies Brand: InfinityLab. LC System Compatibility: 1290 Infinity II LC System. LC Use With Instrument: G7167B. Technique: LC. 1 * 1 Pce
Fournisseur: Agilent
Specifications:- LC Application Compatibility: Analytical. LC Module Type Compatibility: Autosampler Analytical. LC Supplies Brand: InfinityLab. LC System Compatibility: 1290 Infinity II LC System. LC Use With Instrument: G7167B. Technique: LC. 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- LC Application Compatibility: Analytical. LC Module Type Compatibility: Autosampler Analytical. LC Supplies Brand: InfinityLab. LC System Compatibility: 1290 Infinity II LC System. LC Use With Instrument: G7167B. Technique: LC. 1 * 1 Pce HEWL5004-0003 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou