Specifications:- Capillary Material: MP35N. Capillary Subtype: Standard capillary. Fitting Type Left End: SI. Fitting Type Right End : SX. Inner Diameter (ID): 0.17 mm. LC Application Compatibility: Bio-compatible. LC Component Type: Capillary. LC Sy 1 * 1 Pce
Fournisseur: Agilent
Specifications:- Capillary Material: MP35N. Capillary Subtype: Standard capillary. Fitting Type Left End: SI. Fitting Type Right End : SX. Inner Diameter (ID): 0.17 mm. LC Application Compatibility: Bio-compatible. LC Component Type: Capillary. LC Sy 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- Capillary Material: MP35N. Capillary Subtype: Standard capillary. Fitting Type Left End: SI. Fitting Type Right End : SX. Inner Diameter (ID): 0.17 mm. LC Application Compatibility: Bio-compatible. LC Component Type: Capillary. LC Sy 1 * 1 Pce HEWL5004-0041 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou