Specifications:- Capillary Material: MP35N. Capillary Subtype: Standard capillary. Fitting Type Left End: No fitting. Fitting Type R ight End: M4. Inner Diameter (ID): 0.12 mm. LC Application Compatibility: Bio-compatible. LC System Compatibility: 12 1 * 1 Pce
Fournisseur: Agilent
Specifications:- Capillary Material: MP35N. Capillary Subtype: Standard capillary. Fitting Type Left End: No fitting. Fitting Type R ight End: M4. Inner Diameter (ID): 0.12 mm. LC Application Compatibility: Bio-compatible. LC System Compatibility: 12 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- Capillary Material: MP35N. Capillary Subtype: Standard capillary. Fitting Type Left End: No fitting. Fitting Type R ight End: M4. Inner Diameter (ID): 0.12 mm. LC Application Compatibility: Bio-compatible. LC System Compatibility: 12 1 * 1 Pce HEWL5004-0043 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou