Specifications:- Capillary Material: Stainless Steel. Capillary Subtype: Special. Inner Diameter (ID): 0.6 mm. LC Application Compat ibility: Preparative. LC Module Type Compatibility: Fraction Collector. LC System Compatibility: 1290 Infinity II Pur 1 * 1 Pce
Fournisseur: Agilent
Specifications:- Capillary Material: Stainless Steel. Capillary Subtype: Special. Inner Diameter (ID): 0.6 mm. LC Application Compat ibility: Preparative. LC Module Type Compatibility: Fraction Collector. LC System Compatibility: 1290 Infinity II Pur 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- Capillary Material: Stainless Steel. Capillary Subtype: Special. Inner Diameter (ID): 0.6 mm. LC Application Compat ibility: Preparative. LC Module Type Compatibility: Fraction Collector. LC System Compatibility: 1290 Infinity II Pur 1 * 1 Pce HEWL5005-0000 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou