Specifications:- Chemical Compatibility: Organic solvents. Flared: Yes. Inner Diameter (ID): 0.76 mm. Number of Stops: 2. Spectrosco py Instrument Compatibility: 7500 7700 7800 7850 7900 8800 8900 All Agilent ICP-MS systems. Spectroscopy Tubing Mater 1 * 12 Pce
Fournisseur: Agilent
Specifications:- Chemical Compatibility: Organic solvents. Flared: Yes. Inner Diameter (ID): 0.76 mm. Number of Stops: 2. Spectrosco py Instrument Compatibility: 7500 7700 7800 7850 7900 8800 8900 All Agilent ICP-MS systems. Spectroscopy Tubing Mater 1 * 12 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- Chemical Compatibility: Organic solvents. Flared: Yes. Inner Diameter (ID): 0.76 mm. Number of Stops: 2. Spectrosco py Instrument Compatibility: 7500 7700 7800 7850 7900 8800 8900 All Agilent ICP-MS systems. Spectroscopy Tubing Mater 1 * 12 Pce HEWL5005-0026 1 * 12 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou