Provides a typical acquisition time of 210 seconds when used for ICP-MS with ISIS3, peristaltic pump and SPS4. Specifications:- Inne r Diameter (ID): 1 mm. Spectroscopy Instrument Compatibility: 7800 7850 7900 8900. Typical Acquisition Time: 210 sec. 1 * 1 Pce
Fournisseur: Agilent
Provides a typical acquisition time of 210 seconds when used for ICP-MS with ISIS3, peristaltic pump and SPS4. Specifications:- Inne r Diameter (ID): 1 mm. Spectroscopy Instrument Compatibility: 7800 7850 7900 8900. Typical Acquisition Time: 210 sec. 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Provides a typical acquisition time of 210 seconds when used for ICP-MS with ISIS3, peristaltic pump and SPS4. Specifications:- Inne r Diameter (ID): 1 mm. Spectroscopy Instrument Compatibility: 7800 7850 7900 8900. Typical Acquisition Time: 210 sec. 1 * 1 Pce HEWL5005-0036 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou