Provides a typical acquisition time of 130 seconds when used for ICP-MS with ISIS3, peristaltic pump and SPS4. Standard loop supplie d with ISIS3 preconfigured kit (G8411-68202). Specifications:- Inner Diameter (ID): 2.18 mm. Spectroscopy Instrument 1 * 1 Pce
Fournisseur: Agilent
Provides a typical acquisition time of 130 seconds when used for ICP-MS with ISIS3, peristaltic pump and SPS4. Standard loop supplie d with ISIS3 preconfigured kit (G8411-68202). Specifications:- Inner Diameter (ID): 2.18 mm. Spectroscopy Instrument 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Provides a typical acquisition time of 130 seconds when used for ICP-MS with ISIS3, peristaltic pump and SPS4. Standard loop supplie d with ISIS3 preconfigured kit (G8411-68202). Specifications:- Inner Diameter (ID): 2.18 mm. Spectroscopy Instrument 1 * 1 Pce HEWL5005-0040 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou